siRNA Details
| Gene Symbol: Zbtb11os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027965.1 | 1 | 1292 | |||
| GenBank mRNA | AK006543.1 | - | 1 | 1090 | ||
| AK010004.1 | - | 1 | 1267 | |||
| BC017620.1 | - | 1 | 1226 | 4924337 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb11os1| Gene Symbol: Zbtb11os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027965.1 | 1 | 1292 | |||
| GenBank mRNA | AK006543.1 | - | 1 | 1090 | ||
| AK010004.1 | - | 1 | 1267 | |||
| BC017620.1 | - | 1 | 1226 | 4924337 | ||