siRNA Details
| Gene Symbol: Gm5477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002869.1 | 3 | 479 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5477| Gene Symbol: Gm5477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002869.1 | 3 | 479 | |||