siRNA Details
| Gene Symbol: Cyp2d13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003552.1 | 3 | 472 | |||
| GenBank mRNA | AK004933.1 | - | 302 | |||
| BC018344.1 | - | 3 | 452 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2d13| Gene Symbol: Cyp2d13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003552.1 | 3 | 472 | |||
| GenBank mRNA | AK004933.1 | - | 302 | |||
| BC018344.1 | - | 3 | 452 | |||