siRNA Details
| Gene Symbol: Cyp2d13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003552.1 | 5, 6 | 918 | |||
| GenBank mRNA | AK004933.1 | - | 748 | |||
| BC018344.1 | - | 5, 6 | 898 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI147110.1 | - | 2, 3 | 340 | Chr. 15 - 82636826 - 82640267 [-] on Build GRCm38 | |

Cyp2d13
Genomic Map