siRNA Details
| Gene Symbol: Pvt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003368.1 | 8 | 1243 | |||
| GenBank mRNA | AK020628.1 | - | 6 | 974 | ||
| AK169445.1 | - | 8 | 1184 | |||
| BC027817.1 | - | 3 | 311 | |||
| BC049751.1 | - | 4 | 491 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pvt1| Gene Symbol: Pvt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003368.1 | 8 | 1243 | |||
| GenBank mRNA | AK020628.1 | - | 6 | 974 | ||
| AK169445.1 | - | 8 | 1184 | |||
| BC027817.1 | - | 3 | 311 | |||
| BC049751.1 | - | 4 | 491 | |||