siRNA Details
Gene Symbol: 4833418N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015506.2 | 2, 3 | 853 | |||
GenBank mRNA | AK031641.1 | - | 2, 3 | 856 | ||
AK082147.1 | - | 1, 2 | 1141 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4833418N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015506.2 | 2, 3 | 853 | |||
GenBank mRNA | AK031641.1 | - | 2, 3 | 856 | ||
AK082147.1 | - | 1, 2 | 1141 |