siRNA Details
| Gene Symbol: 4833418N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015506.2 | 5 | 1325 | |||
| GenBank mRNA | AK031641.1 | - | 5 | 1328 | ||
| AK082147.1 | - | 4 | 1613 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833418N02Rik| Gene Symbol: 4833418N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015506.2 | 5 | 1325 | |||
| GenBank mRNA | AK031641.1 | - | 5 | 1328 | ||
| AK082147.1 | - | 4 | 1613 | |||