siRNA Details
Gene Symbol: 4833418N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015506.2 | 2 | 704 | |||
GenBank mRNA | AK014719.1 | - | 2 | 929 | ||
AK028291.1 | - | 1 | 2081 | |||
AK031641.1 | - | 2 | 707 | |||
AK082147.1 | - | 1 | 992 |
