siRNA Details
Gene Symbol: 4833418N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015506.2 | 5 | 1278 | |||
GenBank mRNA | AK031641.1 | - | 5 | 1281 | ||
AK082147.1 | - | 4 | 1566 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4833418N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015506.2 | 5 | 1278 | |||
GenBank mRNA | AK031641.1 | - | 5 | 1281 | ||
AK082147.1 | - | 4 | 1566 |