siRNA Details
Gene Symbol: 4833418N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015506.2 | 1 | 452 | |||
GenBank mRNA | AK014719.1 | - | 1 | 418 | ||
AK028291.1 | - | 1 | 417 | |||
AK031641.1 | - | 1 | 455 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4833418N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015506.2 | 1 | 452 | |||
GenBank mRNA | AK014719.1 | - | 1 | 418 | ||
AK028291.1 | - | 1 | 417 | |||
AK031641.1 | - | 1 | 455 |