siRNA Details
| Gene Symbol: C030016D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027987.1 | 1 | 183 | |||
| GenBank mRNA | AK047713.1 | - | 1 | 149 | ||
| BC094907.1 | - | 1 | 183 | |||
| BQ559386.1 | - | 1 | 183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C030016D13Rik| Gene Symbol: C030016D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027987.1 | 1 | 183 | |||
| GenBank mRNA | AK047713.1 | - | 1 | 149 | ||
| BC094907.1 | - | 1 | 183 | |||
| BQ559386.1 | - | 1 | 183 | |||