siRNA Details
| Gene Symbol: Gm10319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003624.2 | 2 | 348 | |||
| GenBank mRNA | BC061166.1 | - | 2 | 348 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA209587.1 | - | 1 | 102 | Chr. 6 - 122138375 - 122139805 [+] on Build GRCm38 | |

Gm10319
Genomic Map