siRNA Details
| Gene Symbol: A430093F15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027805.1 | 4 | 1890 | |||
| GenBank mRNA | AK079853.1 | - | 4 | 1894 | ||
| BC064125.1 | - | 4 | 769 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_207529.1 | - | 1894 | Chr. - - [-] on Build | ||

A430093F15Rik
Genomic Map