siRNA Details
| Gene Symbol: Gm4566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028023.1 | 3 | 514 | |||
| GenBank mRNA | DV658461.1 | - | 2 | 636 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4566| Gene Symbol: Gm4566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028023.1 | 3 | 514 | |||
| GenBank mRNA | DV658461.1 | - | 2 | 636 | ||