siRNA Details
| Gene Symbol: Gm4566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028023.1 | 3 | 412 | |||
| GenBank mRNA | CO802078.1 | - | 3 | 414 | ||
| DV658461.1 | - | 2 | 534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4566| Gene Symbol: Gm4566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028023.1 | 3 | 412 | |||
| GenBank mRNA | CO802078.1 | - | 3 | 414 | ||
| DV658461.1 | - | 2 | 534 | |||