siRNA Details
| Gene Symbol: 1700073E17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003625.1 | 1 | 762 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700073E17Rik| Gene Symbol: 1700073E17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003625.1 | 1 | 762 | |||