siRNA Details
Gene Symbol: 1700073E17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003625.1 | 1 | 4510 | |||
GenBank mRNA | BC049631.1 | - | 2 | 982 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB679532.1 | - | 1 | 323 | Chr. 6 - 145391812 - 145392230 [+] on Build GRCm38 |
