siRNA Details
| Gene Symbol: Snora31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028481.1 | 1 | 46 | |||
| GenBank mRNA | AF357400.1 | - | 1 | 46 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA120090.1 | - | 1 | 292 | Chr. 14 - 75847677 - 75848278 [+] on Build GRCm38 | |

Snora31
Genomic Map