siRNA Details
| Gene Symbol: Snora31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028481.1 | 1 | 99 | |||
| GenBank mRNA | AF357400.1 | - | 1 | 99 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snora31| Gene Symbol: Snora31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028481.1 | 1 | 99 | |||
| GenBank mRNA | AF357400.1 | - | 1 | 99 | ||