siRNA Details
| Gene Symbol: A730017L22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015523.2 | 3 | 698 | |||
| GenBank mRNA | AK158456.1 | - | 3 | 699 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A730017L22Rik| Gene Symbol: A730017L22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015523.2 | 3 | 698 | |||
| GenBank mRNA | AK158456.1 | - | 3 | 699 | ||