siRNA Details
Gene Symbol: A730017L22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015523.2 | 3 | 618 | |||
GenBank mRNA | AK158456.1 | - | 3 | 619 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A730017L22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015523.2 | 3 | 618 | |||
GenBank mRNA | AK158456.1 | - | 3 | 619 |