siRNA Details
| Gene Symbol: Nt5c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028353.1 | 4, 5 | 329 | |||
| GenBank mRNA | AK030786.1 | - | 4, 5 | 287 | ||
| AK181552.1 | - | 2, 3 | 131 | |||
| AK187621.1 | - | 2, 3 | 131 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nt5c2| Gene Symbol: Nt5c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028353.1 | 4, 5 | 329 | |||
| GenBank mRNA | AK030786.1 | - | 4, 5 | 287 | ||
| AK181552.1 | - | 2, 3 | 131 | |||
| AK187621.1 | - | 2, 3 | 131 | |||