siRNA Details
| Gene Symbol: Gm3434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030729.1 | 2, 3 | 203 | |||
| GenBank mRNA | CJ054175.1 | - | 2, 3 | 204 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm3434| Gene Symbol: Gm3434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030729.1 | 2, 3 | 203 | |||
| GenBank mRNA | CJ054175.1 | - | 2, 3 | 204 | ||