siRNA Details
| Gene Symbol: Gm10863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029470.1 | 3 | 295 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF302801.1 | - | 3 | 295 | Chr. 15 - 79166066 - 79216497 [+] on Build GRCm38 | |

Gm10863
Genomic Map