siRNA Details
| Gene Symbol: Gm10863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029470.1 | 5 | 488 | |||
| GenBank mRNA | AK145218.1 | - | 263 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10863| Gene Symbol: Gm10863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029470.1 | 5 | 488 | |||
| GenBank mRNA | AK145218.1 | - | 263 | |||