siRNA Details
Gene Symbol: Hoxaas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015611.2 | 2 | 217 | |||
GenBank mRNA | AK012572.1 | - | 2 | 150 | ||
AK028207.1 | - | 2 | 218 | |||
AK146012.1 | - | 112 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hoxaas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015611.2 | 2 | 217 | |||
GenBank mRNA | AK012572.1 | - | 2 | 150 | ||
AK028207.1 | - | 2 | 218 | |||
AK146012.1 | - | 112 |