siRNA Details
Gene Symbol: Hoxaas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015611.2 | 3 | 617 | |||
GenBank mRNA | AK012572.1 | - | 3 | 550 | ||
AK028207.1 | - | 3 | 618 | |||
AK146012.1 | - | 512 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hoxaas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015611.2 | 3 | 617 | |||
GenBank mRNA | AK012572.1 | - | 3 | 550 | ||
AK028207.1 | - | 3 | 618 | |||
AK146012.1 | - | 512 |