siRNA Details
Gene Symbol: Gm4598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030681.1 | 1 | 109 | |||
GenBank mRNA | AK087307.1 | - | 1 | 110 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB664514.1 | - | 1 | 110 | Chr. 7 - 24661294 - 24662662 [+] on Build GRCm38 |
