siRNA Details
| Gene Symbol: Gm4598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030681.1 | 1 | 160 | |||
| GenBank mRNA | AK087307.1 | - | 1 | 161 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB664514.1 | - | 1 | 161 | Chr. 7 - 24661294 - 24662662 [+] on Build GRCm38 | |

Gm4598
Genomic Map