siRNA Details
| Gene Symbol: BB123696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027893.1 | 3 | 1546 | |||
| GenBank mRNA | AK035379.1 | - | 4 | 1199 | ||
| AK035586.1 | - | 3 | 1539 | |||
| AK035638.1 | - | 2 | 1438 | |||
| AK035711.1 | - | 3 | 1532 | |||
| AK039814.1 | - | 3 | 1548 | |||

BB123696
Genomic Map