siRNA Details
| Gene Symbol: BB123696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027893.1 | 3 | 743 | |||
| GenBank mRNA | AK035379.1 | - | 3 | 729 | ||
| AK035586.1 | - | 3 | 736 | |||
| AK035638.1 | - | 2 | 635 | |||
| AK035711.1 | - | 3 | 729 | |||
| AK039814.1 | - | 3 | 743 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DV051390.1 | - | 3 | 756 | Chr. 13 - 52726197 - 52735591 [-] on Build GRCm38 | |

BB123696
Genomic Map