siRNA Details
| Gene Symbol: BB123696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027893.1 | 3 | 990 | |||
| GenBank mRNA | AK035586.1 | - | 3 | 983 | ||
| AK035638.1 | - | 2 | 882 | |||
| AK035711.1 | - | 3 | 976 | |||
| AK039814.1 | - | 3 | 991 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BB123696| Gene Symbol: BB123696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027893.1 | 3 | 990 | |||
| GenBank mRNA | AK035586.1 | - | 3 | 983 | ||
| AK035638.1 | - | 2 | 882 | |||
| AK035711.1 | - | 3 | 976 | |||
| AK039814.1 | - | 3 | 991 | |||