siRNA Details
| Gene Symbol: Peg3os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023846.1 | 1 | 642 | |||
| GenBank mRNA | EF635411.1 | - | 1 | 642 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Peg3os| Gene Symbol: Peg3os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023846.1 | 1 | 642 | |||
| GenBank mRNA | EF635411.1 | - | 1 | 642 | ||