siRNA Details
| Gene Symbol: Serp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027699.1 | 4 | 377 | |||
| GenBank mRNA | BC100483.1 | - | 4 | 346 | ||
| BC115492.1 | - | 4 | 376 | 40100869 | ||
| BC115493.1 | - | 3 | 303 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serp2| Gene Symbol: Serp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027699.1 | 4 | 377 | |||
| GenBank mRNA | BC100483.1 | - | 4 | 346 | ||
| BC115492.1 | - | 4 | 376 | 40100869 | ||
| BC115493.1 | - | 3 | 303 | |||