siRNA Details
Gene Symbol: 2610001J05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024619.1 | 3 | 1538 | |||
GenBank mRNA | AK035027.1 | - | 3 | 1521 | ||
AK152937.1 | - | 1574 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2610001J05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024619.1 | 3 | 1538 | |||
GenBank mRNA | AK035027.1 | - | 3 | 1521 | ||
AK152937.1 | - | 1574 |