siRNA Details
| Gene Symbol: 2610001J05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024619.1 | 3 | 1199 | |||
| GenBank mRNA | AK035027.1 | - | 3 | 1182 | ||
| AK152937.1 | - | 1235 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610001J05Rik| Gene Symbol: 2610001J05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024619.1 | 3 | 1199 | |||
| GenBank mRNA | AK035027.1 | - | 3 | 1182 | ||
| AK152937.1 | - | 1235 | ||||