siRNA Details
| Gene Symbol: 2810032G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015579.1 | 1115 | ||||
| GenBank mRNA | AK012851.1 | - | 1751 | |||
| AK028030.1 | - | 1635 | ||||
| AK029678.1 | - | 1873 | ||||
| BC065777.1 | - | 1115 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810032G03Rik| Gene Symbol: 2810032G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015579.1 | 1115 | ||||
| GenBank mRNA | AK012851.1 | - | 1751 | |||
| AK028030.1 | - | 1635 | ||||
| AK029678.1 | - | 1873 | ||||
| BC065777.1 | - | 1115 | ||||