siRNA Details
Gene Symbol: 1600029I14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028123.1 | 1 | 123 | |||
GenBank mRNA | AK005560.1 | - | 1 | 103 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1600029I14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028123.1 | 1 | 123 | |||
GenBank mRNA | AK005560.1 | - | 1 | 103 |