siRNA Details
| Gene Symbol: 5730522E02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027973.1 | 6 | 640 | |||
| GenBank mRNA | BY724063.1 | - | 6 | 634 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5730522E02Rik| Gene Symbol: 5730522E02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027973.1 | 6 | 640 | |||
| GenBank mRNA | BY724063.1 | - | 6 | 634 | ||