siRNA Details
Gene Symbol: Otx2os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_029384.1 | 4 | 437 | |||
GenBank mRNA | AK042665.1 | - | LIT3409 Otx2OS |
4 | 437 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Otx2os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_029384.1 | 4 | 437 | |||
GenBank mRNA | AK042665.1 | - | LIT3409 Otx2OS |
4 | 437 |