siRNA Details
| Gene Symbol: Asb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003961.1 | 6 | 1063 | |||
| GenBank mRNA | AK051017.1 | - | 6 | 976 | ||
| BC062948.1 | - | 2 | 217 | 30532932 | ||
| BC066824.1 | - | 4 | 490 | 30087227 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asb7| Gene Symbol: Asb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003961.1 | 6 | 1063 | |||
| GenBank mRNA | AK051017.1 | - | 6 | 976 | ||
| BC062948.1 | - | 2 | 217 | 30532932 | ||
| BC066824.1 | - | 4 | 490 | 30087227 | ||