siRNA Details
| Gene Symbol: Asb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003961.1 | 3, 4 | 634 | |||
| GenBank mRNA | AK051017.1 | - | 3, 4 | 547 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB655391.1 | - | 3, 4 | 544 | Chr. 7 - 66681704 - 66689472 [-] on Build GRCm38 | |

Asb7
Genomic Map