siRNA Details
| Gene Symbol: B430010I23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015457.1 | 3 | 1981 | |||
| GenBank mRNA | AK033372.1 | - | 3 | 1981 | ||
| AK046663.1 | - | 1 | 2385 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B430010I23Rik| Gene Symbol: B430010I23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015457.1 | 3 | 1981 | |||
| GenBank mRNA | AK033372.1 | - | 3 | 1981 | ||
| AK046663.1 | - | 1 | 2385 | |||