siRNA Details
| Gene Symbol: B430010I23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015457.1 | 1 | 168 | |||
| GenBank mRNA | AK033372.1 | - | 1 | 168 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB624594.1 | - | 1 | 168 | Chr. 8 - 41017468 - 41022512 [+] on Build GRCm38 | |

B430010I23Rik
Genomic Map