siRNA Details
Gene Symbol: B430010I23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015457.1 | 3 | 2209 | |||
GenBank mRNA | AK033372.1 | - | 3 | 2209 | ||
AK046663.1 | - | 1 | 2613 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B430010I23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015457.1 | 3 | 2209 | |||
GenBank mRNA | AK033372.1 | - | 3 | 2209 | ||
AK046663.1 | - | 1 | 2613 |