siRNA Details
| Gene Symbol: Obox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC094912.1 | - | 3 | 428 | ||
| CA556780.1 | - | 3 | 448 | |||
| DV663188.1 | - | 3 | 547 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Obox2| Gene Symbol: Obox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC094912.1 | - | 3 | 428 | ||
| CA556780.1 | - | 3 | 448 | |||
| DV663188.1 | - | 3 | 547 | |||