siRNA Details
| Gene Symbol: Gm14207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030683.1 | 1, 2 | 213 | |||
| GenBank mRNA | AK039958.1 | - | 1, 2 | 214 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14207| Gene Symbol: Gm14207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030683.1 | 1, 2 | 213 | |||
| GenBank mRNA | AK039958.1 | - | 1, 2 | 214 | ||