siRNA Details
Gene Symbol: Vps39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027618.1 | 3 | 342 | |||
GenBank mRNA | AK170831.1 | - | 3 | 332 | ||
BB655861.1 | - | 3 | 298 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vps39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027618.1 | 3 | 342 | |||
GenBank mRNA | AK170831.1 | - | 3 | 332 | ||
BB655861.1 | - | 3 | 298 |