siRNA Details
| Gene Symbol: Ftx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028380.1 | 5 | 835 | |||
| NR_028381.1 | 5 | 856 | ||||
| GenBank mRNA | AK035525.1 | - | 5 | 858 | ||
| AK085125.1 | - | 5 | 723 | |||
| BC027389.1 | - | 4 | 631 | |||
| BC076616.1 | - | 5 | 744 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG920590.1 | - | 4 | 635 | Chr. X - 103606955 - 103615193 [-] on Build GRCm38 | |

Ftx
Genomic Map