siRNA Details
| Gene Symbol: Ftx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028380.1 | 6, 7 | 961 | |||
| NR_028381.1 | 6, 7 | 982 | ||||
| GenBank mRNA | AK035525.1 | - | 6, 7 | 984 | ||
| AK085125.1 | - | 6, 7 | 849 | |||
| BC027389.1 | - | 5, 6 | 757 | |||
| BC076616.1 | - | 6, 7 | 870 | |||

Ftx
Genomic Map